Learn More
NKX2-2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NKX2-2.
Marca: Abnova PAB25798.100ug
Descrizione
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor. [provided by RefSeq
Specifica
| NKX2-2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| NKX2-2 | |
| NKX2-2 | |
| A synthetic peptide corresponding to 19 amino acids at internal region of human NKX2-2. | |
| 100 μg | |
| Primary | |
| NKX2-2 antibody is predicted to not cross-react with other NK2 homeobox family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of NKX2-2. | |
| In PBS (0.02% sodium azide) | |
| NKX2.2/NKX2B | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 4821 | |
| Human, Mouse, Rat | |
| Liquid |
For Research Use Only