Learn More
PAK1 (phospho T423), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK1.
Marca: Abnova PAB8092.400uL
Descrizione
PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifica
| PAK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK1. | |
| In PBS (0.09% sodium azide) | |
| MGC130000/MGC130001/PAKalpha | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 5058 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA | |
| Unconjugated | |
| ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| PAK1 | |
| PAK1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T423 of human PAK1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only