Learn More
Grant Instruments™ Raised Shelf for Unstirred Water Baths
Codice prodotto. 11416487
Click to view available options
Tipo:
Shelf for 12 L Tanks
Shelf for 18 L Tanks
Shelf for 26 L Tanks
Shelf for 34 L Tanks
Dimensione della confezione:
Pezzo
Codice prodotto. 11416487
Marca: Grant Instruments™ RS28
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (apparecchiatura) Grant Heated Circulating Baths
Tipo Shelf for 26 L Tanks
Altezza (sistema metrico) 45 or 135 mm