missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgE Heavy Chain Secondary Antibody (LO-ME-3), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NBP2-68431IR
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgE Heavy Chain Monoclonal antibody specifically detects IgE Heavy Chain in Mouse samples. It is validated for ELISA, Western Blot (Negative)
Specifica
| IgE Heavy Chain | |
| Monoclonal | |
| DyLight 755 | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Purified IgE from BALB/c mice | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot (Negative) | |
| LO-ME-3 | |
| 50mM Sodium Borate | |
| Rat | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction