missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Anti-Mouse IgG2A PerCP-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems F0131
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Flow Cytometry.
Specifica
| IgG2a | |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cocktail of mouse IgG 1 and IgG 2A | |
| 100 Tests | |
| Secondary | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 344701 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Rat | |
| Affinity Purified | |
| RUO | |
| The specificity of this antibody was confirmed by direct ELISA on mouse IgG 2A monoclonal antibodies, and it does not cross-react with IgG 1 IgG 2B IgG 3 IgM, IgA, or IgE antibodies. By flow cytometry, this isotype-specific secondary antibody detects primary antibodies of the mouse IgG 2A isotype, and does not detect primary antibodies of mouse IgG 1 IgG 2B or IgG 3 class. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto