missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat C-Reactive Protein/CRP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB17441-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifica
| C-Reactive Protein/CRP | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P48199 | |
| CRP | |
| Mouse myeloma cell line NS0-derived recombinant rat C-Reactive Protein/CRP His20-Ser230 Accession # P48199 | |
| 25 μg | |
| Primary | |
| Detects rat C-Reactive Protein/CRP in direct ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse CRP, recombinant porcine CRP, recombinant human (rh) Pentraxin 2, or rhPentraxin 3 is observed. | |
| Rat | |
| Purified |
| ELISA | |
| 233539 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1401 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto