Learn More
R&D Systems™ Rat C-Reactive Protein/CRP DuoSet ELISA
Codice prodotto. 30021605
Click to view available options
Quantità:
15 x 96-well Plates
Dimensione della confezione:
1 set
Codice prodotto. 30021605
Marca: R&D Systems™ DY1744
Rat C-Reactive Protein/CRP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Rat C-Reactive Protein/CRP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies rat C-Reactive Protein/CRP.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 15 x 96-well Plates
Coniugato Biotin
Intervallo saggio 156.00 to 10000 pg/mL
Da utilizzare con (applicazione) ELISA
ID gene (immissione) 25419
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.