Learn More
RBPJ Mouse anti-Human, Clone: OTI4B11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF809493
Descrizione
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.Specifica
| RBPJ | |
| Monoclonal | |
| Unconjugated | |
| RBPJ | |
| AOS3, CBF1, IGKJRB, IGKJRB1, KBF2, RBP-J, RBPJK, RBPSUH, SUH, csl | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3516 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4B11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q06330 | |
| RBPJ | |
| Human recombit protein fragment corresponding to amino acids 1-243 of human RBPJ produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.