Learn More
Thermo Scientific™ Reacti-Therm™ Thermometers
Codice prodotto. 11537510 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Temperatura di esercizio (sistema metrico):
0°C to 100°C
0°C to 200°C
Dimensione della confezione:
Pezzo
Codice prodotto. 11537510
Marca: Thermo Scientific™ TS18914
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use these thermometers to measure temperature in dry block heaters of Thermo Scientific™ Reacti-Therm™ Heating and Stirring module.
Measure temperature in dry block heaters of the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module and other laboratory equipment using Thermo Scientific™ Reacti-Therm™ Thermometers. The thermometers are mercury-free and alcohol-filled for greater safety. PTFE coating ensures that glass is impervious to corrosive materials. They are available in two different temperature ranges.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Thermometer
Temperatura di esercizio (sistema metrico) 0°C to 100°C
Da utilizzare con (apparecchiatura) Dry Block Heaters and Other Instruments