Learn More
Lenz Laborglasintrumente Reaction Vessels, Cylindrical, with Thermostatic Jacket and Withdrawal Valve
Click to view available options
Altezza (sistema metrico):
245 mm
275 mm
Include:
Connector GL 14
Connector KF 15
Dimensione della confezione:
Pezzo
Descrizione
- With graduation
- With laboratory flange LF
- With groove
- With two connectors (with threads GL 14 /GL 18 or ball flanges KF 15 / KF 25)
- Made of DURAN™ tubing
- Other sizes on request
- Delivery with 2 plastic olives GL14/18 and screw caps
Specifica
Specifica
| Capacità (sistema metrico) | 0.1 L |
| Materiale | Duran Glass |
| Altezza (sistema metrico) | 245 mm |
| Include | Connector GL 14 |