Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket
Click to view available options
Diametro (sistema metrico) esterno:
105 mm
131 mm
165 mm
206 mm
236 mm
280 mm
350 mm
Altezza (sistema metrico):
155 mm
180 mm
215 mm
260 mm
295 mm
340 mm
355 mm
420 mm
Include:
Lab Flange 100 LF, Connector GL 18
Lab Flange 100 LF, Connector KF 15
Lab Flange 150 LF, Connector GL 18
Lab Flange 200 LF, Connector KF 15
Lab Flange 60 LF, Connector GL 14
Lab Flange 60 LF, Connector KF 15
Dimensione della confezione:
Pezzo
Descrizione
- Without graduation
- With laboratory flange LF
- With groove, with two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
- Made of DURAN™ tubing
- Other sizes on request
Specifica
Specifica
| Capacità (sistema metrico) | 2 L |
| Materiale | Duran Glass |
| Altezza (sistema metrico) | 260 mm |
| Include | Lab Flange 100 LF, Connector GL 18 |
| Diametro (sistema metrico) esterno | 206 mm |