Learn More
BD Biosciences Reagent Rack - FITC
Click to view available options
:
Holds: 5 BD™ Amber vials and 20 narrower BD Pharmingen™ type vials.
Descrizione
Pre-cooled BD™ Reagent Racks keep your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability. The BD Reagent Racks are stackable and available in different colors to help you keep your vials organized on the bench as well as in the fridge. Each rack is filled with cooling material, and can accommodate up to 25 BD™ Amber vials and 20 narrower BD Pharmingen™ type vials. Size 180 mm x 54,5 mm - Weight: 950 gr.
Specifica
Specifica
| Include | Filled with cooling material. |
| Capacità | BD Amber Vials (5) and Narrower BD Pharmingen Type Vials (20) |
| Sovrapponibile | Sì |
| Dimensioni (L x P) | 180 mm x 54.5 mm |
| Peso (sistema metrico) | 950 G |