Learn More
enQuireBio™ Recombinant C. trachomatis Major outer membrane porin Protein
Codice prodotto. 16078030
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Codice prodotto. 16078030
Marca: enQuireBio™ QP7292ec1mg
A DNA sequence encoding the Chlamydia trachomatis serovar L2 (strain 434/Bu / ATCC VR-902B) Major outer membrane porin, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso WP_009873292.1
ID gene (immissione) 5858320
Nome Major outer membrane porin Protein
Quantità 1 mg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene ompA
Lunghezza della proteina Full Length
Product Type Recombinant Protein
Cross-reattività C. trachomatis
Specie E. coli
Tag proteine His-SUMO
Sequenza LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLQTDVNKEF QMGAKPTTAT GNAAAPSTCT ARENPAYGRH MQDAEMFTNA AYMALNIWDR FDVFCTLGAT SGYLKGNSAS FNLVGLFGDN ENHATVSDSK LVPNMSLDQS VVELYTDTTF AWSAGARAAL WECGCATLGA SFQYAQSKPK VEELNVLCNA AEFTINKPKG YVGQEFPLDL KAGTDGVTGT KDASIDYHEW QASLALSYRL NMFTPYIGVK WSRASFDADT IRIAQPKSAT TVFDVTTLNP TIAGAGDVKA SAEGQLGDTM QIVSLQLNKM KSRKSCGIAV GTTIVDADKY AVTVETRLID ERAAHVNAQF RF
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.