missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Drosophila Decapentaplegic (Dpp) Protein
Click to view available options
Formulazione:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Dimensione della confezione:
20µg
Descrizione
The Recombinant Drosophila Decapentaplegic (Dpp) Protein is derived from E. coli. The Recombinant Drosophila Decapentaplegic (Dpp) Protein has been validated for the following applications: Bioactivity.
Specifica
Specifica
| Formulazione | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Peso molecolare | 14.8 kDa (monomer) |
| Nome | Decapentaplegic/DPP |
| Quantità | 20 μg |
| Sorgente | E. coli-derived drosophila Decapentaplegic/DPP protein Asp457-Arg588 (Gln473His) & (Pro474Ala),with an N-terminal Met |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Forma strutturale | Disulfide-linked homodimer |
| Coniugato | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto