Learn More
enQuireBio™ Recombinant E. coli Outer-membrane lipoprotein carrier Protein
Codice prodotto. 16088850
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Codice prodotto. 16088850
Marca: enQuireBio™ QP7432ye500ug
A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) Outer-membrane lipoprotein carrier protein, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso WP_001295343.1
Nome Outer-membrane lipoprotein carrier Protein
Quantità 500 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene lolA
Lunghezza della proteina Full Length
Product Type Recombinant Protein
Cross-reattività E. coli
Specie Yeast
Tag proteine His
Sequenza DAASDLKSRL DKVSSFHASF TQKVTDGSGA AVQEGQGDLW VKRPNLFNWH MTQPDESILV SDGKTLWFYN PFVEQATATW LKDATGNTPF MLIARNQSSD WQQYNIKQNG DDFVLTPKAS NGNLKQFTIN VGRDGTIHQF SAVEQDDQRS SYQLKSQQNG AVDAAKFTFT PPQGVTVDDQ RK
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.