Learn More
enQuireBio™ Recombinant Human ATP1B1 Protein
Codice prodotto. 15996148
Click to view available options
Quantità:
1 mg
10 μg
2 μg
20 μg
5 μg
Dimensione della confezione:
10µg
1mg
20µg
2µg
5µg
Codice prodotto. 15996148
Marca: enQuireBio™ QP11104HIS2ug
A cDNA sequence encoding the ATP1B1 was constructed and used to recombinantly synthesize the protein.
Specifica
Nome ATP1B1 Protein
Quantità 2 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His
Sequenza MGSSHHHHHH SSGLVPRGSH MGSEFKPTYQ DRVAPPGLTQ IPQIQKTEIS FRPNDPKSYE AYVLNIVRFL EKYKDSAQRD DMIFEDCGDV PSEPKERGDF NHERGERKVC RFKLEWLGNC SGLNDETYGY KEGKPCIIIK LNRVLGFKPK PPKNESLETY PVMKYNPNVL PVQCTGKRDE DKDKVGNVEY FGLGNSPGFP LQYYPYYGKL LQPKYLQPLL AVQFTNLTMD TEIRIECKAY GENIGYSEKD RFQGRFDVKI EVKS
Tampone The ATP1B1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.