Learn More
enQuireBio™ Recombinant Human BMP-2 Protein
Product Code. p-7152077
missing translation for 'orderingAttributeHoverText'
Quantità:
1 mg
10 μg
100 μg
2 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
2µg
500µg
Product Code. 15995979
missing translation for 'mfr': enQuireBio™ QP53632ug
Recombinant Protein
Specifications
Numero di accesso NP_001191.1
ID gene (immissione) 650
Nome BMP-2 Protein
Quantità 2 μg
Status giuridico Research Use Only
Concentrazione di endotossine Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Simbolo del gene BMP2
Attività biologica E. coli synthesized BMP-2Shows bioactivity at a concentration of 250 ng/mL in the Alkaline phosphatase activity induced in ATDC-5 cells assay. The bioactivity acceptance criteria is set at 4 x 10^3 units/mg. The more potent HEK293 derived BMP2 has a specific activity of less than 40 ng/ml in ATDC-5 cell induced alk phos activity.
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine Untagged
Sequenza MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR
Tampone Lyophilized from a sterile filtered (0.22 um) 2x PBS solution.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.