missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD23/Fc epsilon RII Protein
missing translation for 'orderingAttributeHoverText'
Quantità:
50 μg
Förpackningsstorlek
50µg
Beskrivning
The Recombinant Human CD23/Fc epsilon RII Protein is derived from NS0. The Recombinant Human CD23/Fc epsilon RII Protein has been validated for the following applications: Binding Activity.
Specifikationer
Specifikationer
| Formulazione | Lyophilized from a 0.2μm filtered solution in PBS. |
| ID gene (immissione) | 2208 |
| Peso molecolare | 32 kDa |
| Nome | CD23/Fc epsilon RII |
| Quantità | 50 μg |
| Sorgente | Mouse myeloma cell line,NS0-derived human CD23/Fc epsilon RII protein Asp48-Ser321,with an N-terminal 9-His tag |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Coniugato | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering