Learn More
R&D Systems™ Recombinant Human Dopa Decarboxylase/DDC Protein
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Recombinant Human Dopa Decarboxylase/DDC Protein is derived from Sf 21 (baculovirus). The Recombinant Human Dopa Decarboxylase/DDC Protein has been validated for the following applications: Enzyme Activity.
Specifica
Specifica
| Formulazione | Supplied as a 0.2μm filtered solution in MES, NaCl and DTT. |
| ID gene (immissione) | 1644 |
| Peso molecolare | 55 kDa |
| Nome | Dopa Decarboxylase/DDC |
| Quantità | 10 μg |
| Sorgente | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Dopa Decarboxylase/DDC protein Met1-Glu480,with a C-terminal 6-His tag |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Coniugato | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |