missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human ERK3/MAPK6 GST (N-Term) Protein
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
10 μg
25 μg
Dimensione della confezione:
10µg
25µg
Descrizione
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifica
Specifica
| Da utilizzare con (applicazione) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulazione | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID gene (immissione) | 5597 |
| Peso molecolare | MolecularWeight-theroretical: 109.1 kDa |
| Quantità | 25 μg |
| Requisiti di stoccaggio | Store at -80°C. Avoid freeze-thaw cycles. |
| Alias gene | DKFZp686F03189, EC 2.7.11, EC 2.7.11.24, ERK-3, ERK3p97-MAPK, Extracellular signal-regulated kinase 3, extracellular signal-regulated kinase, p97, HsT17250, MAP kinase 6, MAP kinase isoform p97, MAPK 6, mitogen-activated protein kinase 6, p97MAPK, PRKM6, protein kinase, mitogen-activated 5, protein kinase, mitogen-activated 6 |
| Simbolo del gene | MAPK6 |
| Research Category | Breast Cancer, Cancer, Cell Cycle and Replication, Hypoxia, Signal Transduction |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto