Learn More
R&D Systems™ Recombinant Human FABP3/H-FABP Protein CF
R&D Systems™ Recombinant Human FABP3/H-FABP Protein CF is a member of a large superfamily of lipid binding proteins that are expressed in a tissue specific manner.
Marca: R&D Systems™ 9479-CL-100
Descrizione
The Recombinant Human FABP3/H-FABP Protein is derived from E. coli.Specifica
| P05413 | |
| Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT. | |
| 16kDa | |
| E. coli-derived human FABP3/H-FABP protein, (N-terminus) MHHHHHH GS Human FABP3/H-FABP (Met1-Ala133) Accession &Num; P05413 (C-terminus) | |
| <0.10EU per 1μg of the protein by the LAL method. | |
| Recombinant Protein | |
| Reconstitute at 1mg/mL in PBS. |
| Bioactivity not tested | |
| 2170 | |
| 100μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, HFABP, H-FABP, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
| Unconjugated |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.