Learn More
R&D Systems™ Recombinant Human GST-Ubiquitin E1 Enzyme (UBE1) Protein
Click to view available options
Quantità:
50 μg
Packungsgröße:
50µg
Beschreibung
Recombinant Human GST-Ubiquitin E1 Enzyme (UBE1) is a member of the Ubiquitin-activating (E1) enzyme family that is required for the first step of the enzymatic cascade that subsequently utilizes a Ubiquitin-conjugating (E2) enzyme and a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human GST-Ubiquitin E1 Enzyme (UBE1) concentration of 50-200 nM.
Spezifikation
Spezifikation
| Numero di accesso | P22314.3 |
| Da utilizzare con (applicazione) | Enzyme Activity |
| Formulazione | Supplied as a 0.2 μm filtered solution in HEPES & NaCl. |
| ID gene (immissione) | 7317 |
| Peso molecolare | M.W. (theoretical): 148 kDa |
| Nome | GST Ubiquitin-activating Enzyme/UBE1 |
| Quantità | 50 μg |
| Sorgente | Spodoptera frugiperda, Sf 9 (baculovirus)-derived human Ubiquitin-activating Enzyme/UBE1 protein Met1 - Arg1058 with a N-terminal GST (glutathione S-transferase) tag |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Simbolo del gene | UBA1 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur