missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IGF-I/IGF-1 LR3 Protein
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
1 mg
5 mg
20 mg
Dimensione della confezione:
1mg
20mg
5mg
Descrizione
Assay 1: Fully biologically active when compared to standard. Measured in a serum-free cell proliferation assay using human MCF-7 cells. The ED50 for this effect is 0.3-1.5 ng/mL, corresponding to a specific activity of > 6.7 x 10^5 U/mg. Assay 2: Fully biologically active when compared to standard. The ED50 as determined by the stimulation of protein synthesis using rat L6 myoblasts is less than 10 ng/mL, corresponding to a specific activity of > 1.0 x 10^5 U/mg.
Specifica
Specifica
| Da utilizzare con (applicazione) | SDS-PAGE, Bioactivity |
| Formulazione | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.2. |
| ID gene (immissione) | 3479 |
| Peso molecolare | MolecularWeight-theroretical: 9.1 kDa |
| Quantità | 1 mg |
| Requisiti di stoccaggio | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Concentrazione di endotossine | Less than 0.01 EU/μg of IGF-I/IGF-1 AS determined by LAL method. |
| Alias gene | IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C |
| Simbolo del gene | IGF1 |
| Ricostituzione | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto