missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interleukin-21 / IL-21 Protein
Recombinant Protein
Marca: enQuireBio™ QP5394-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001193935.1 | |
Interleukin-21 / IL-21 Protein | |
Research Use Only | |
IL21 | |
Recombinant Protein | |
E. coli | |
MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE | |
Greater than 95% as determined by SDS-PAGE. |
59067 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Measured by mino cell proliferation. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |