Learn More
enQuireBio™ Recombinant Human Laminin subunit beta-2 Protein
Codice prodotto. 16086461
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Codice prodotto. 16086461
Marca: enQuireBio™ QP8788ec100ug
A DNA sequence encoding the Homo sapiens (Human) Laminin subunit beta-2, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso NP_002283.3
ID gene (immissione) 3913
Nome Laminin subunit beta-2 Protein
Quantità 100 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene LAMB2
Lunghezza della proteina Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His
Sequenza PDSIEMVATR VLELSIPASA EQIQHLAGAI AERVRSLADV DAILARTVGD VRRAEQLLQD ARRARSWAED EKQKAETVQA ALEEAQRAQG IAQGAIRGAV ADTRDTEQTL YQVQERMAGA ERALSSAGER ARQLDALLEA LKLKRAGNSL AASTAEETAG SAQGRAQEAE QLLRGPLGDQ YQTVKALAER KAQGVLAAQA RAEQLRDEAR DLLQAAQDKL QRLQELEGTY EENERALESK AAQLDGLEAR MRSVLQAINL QVQIYNTCQ
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.