Learn More
enQuireBio™ Recombinant Human LRG1 Protein
Codice prodotto. p-7150454
Click to view available options
Quantità:
1 mg
10 μg
2 μg
Dimensione della confezione:
10µg
1mg
2µg
Codice prodotto. 15910899
Marca: enQuireBio™ QP125832ug
A cDNA sequence encoding the LRG1 was constructed and used to recombinantly synthesize the protein.
Specifica
Nome LRG1 Protein
Quantità 2 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Human
Specie Human
Tag proteine Untagged
Sequenza VTLSPKDCQV FRSDHGSSIS CQPPAEIPGY LPADTVHLAV EFFNLTHLPA NLLQGASKLQ ELHLSSNGLE SLSPEFLRPV PQLRVLDLTR NALTGLPPGL FQASATLDTL VLKENQLEVL EVSWLHGLKA LGHLDLSGNR LRKLPPGLLA NFTLLRTLDL GENQLETLPP DLLRGPLQLE RLHLEGNKLQ VLGKDLLLPQ PDLRYLFLNG NKLARVAAGA FQGLRQLDML DLSNNSLASVPEGLWASLGQ PNWDMRDGFD ISGNPWICDQ NLSDLYRWLQ AQKDKMFSQN DTRCAGPEAV KGQTLLAVAK SQ
Tampone LRG1 protein filtered (0.4microm) and lyophilized from 20mM Tris and 20mM NaCl, pH 8.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.