missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MBL-2 / MBL Protein
A DNA sequence encoding the Homo sapiens (Human) MBL-2 / MBL, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9268-ye-100ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_000233.1 | |
MBL-2 / MBL Protein | |
Research Use Only | |
MBL2 | |
Recombinant Protein | |
Yeast | |
ETVTCEDAQK TCPAVIACSS PGINGFPGKD GRDGTKGEKG EPGQGLRGLQ GPPGKLGPPG NPGPSGSPGP KGQKGDPGKS PDGDSSLAAS ERKALQTEMA RIKKWLTFSL GKQVGNKFFL TNGEIMTFEK VKALCVKFQA SVATPRNAAE NGAIQNLIKE EAFLGITDEK TEGQFVDLTG NRLTYTNWNE GEPNNAGSDE DCVLLLKNGQ WNDVPCSTSH LAVCEFPI | |
Greater than 90% as determined by SDS-PAGE. |
4153 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |