Learn More
R&D Systems™ Recombinant Human PA28 Activator alpha Subunit Protein
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
Descrizione
The Recombinant Human PA28 Activator alpha Subunit Protein is derived from E. coli. The Recombinant Human PA28 Activator alpha Subunit Protein has been validated for the following applications: Bioactivity.
Specifica
Specifica
| Quantità | 100 μg |
| Specie | Human |
| Content And Storage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |