missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PEX26 Protein
A cDNA sequence encoding the PEX26 was constructed and used to recombinantly synthesize the protein.
120.00€ - 4795.00€
Specifica
Nome | PEX26 Protein |
---|---|
Purezza | Greater than 90% as determined by SDS-PAGE. |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15902249
|
enQuireBio™
QP13012-5UG |
5 μg |
120.00€
5µg |
Eseguire il login per visualizzare lo stock disponibile |
15992239
|
enQuireBio™
QP13012-20UG |
20 μg |
181.00€
20µg |
Eseguire il login per visualizzare lo stock disponibile |
15982239
|
enQuireBio™
QP13012-1MG |
1 mg |
4795.00€
1mg |
Eseguire il login per visualizzare lo stock disponibile |
Specifica
PEX26 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKSDSST SAAPLRGLGG PLRSSEPVRA VPARAPAVDL LEEAADLLVV HLDFRAALET CERAWQSLAN HAVAEEPAGT SLEVKCSLCV VGIQALAEMD RWQEVLSWVL QYYQVPEKLP PKVLELCILL YSKMQEPGAV LDVVGAWLQD PANQNLPEYG ALAEFHVQRV LLPLGCLSEA EELVVGSAAF GEERRLDVLQ AIHTARQQQK QEHSGSEEAQ KPNLEGSVSH KFLSLPMLVR QLWDSAVSH |
Greater than 90% as determined by SDS-PAGE. | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PEX26 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |