Learn More
enQuireBio™ Recombinant Human PGP Protein
Codice prodotto. 15945258
Click to view available options
Quantità:
1 mg
10 μg
2 μg
Dimensione della confezione:
10µg
1mg
2µg
Codice prodotto. 15945258
Marca: enQuireBio™ QP108162ug
A cDNA sequence encoding the PGP was constructed and used to recombinantly synthesize the protein.
Specifica
Nome PGP Protein
Quantità 2 μg
Status giuridico Research Use Only
Concentrazione di endotossine < 1.0 EU per ug protein as determined by the LAL method.
Attività biologica Specific activity is > 3,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His
Sequenza MGSSHHHHHH SSGLVPRGSH MGSHMAAAEA GGDDARCVRL SAERAQALLA DVDTLLFDCD GVLWRGETAV PGAPEALRAL RARGKRLGFI TNNSSKTRAA YAEKLRRLGF GGPAGPGASL EVFGTAYCTA LYLRQRLAGA PAPKAYVLGS PALAAELEAV GVASVGVGPE PLQGEGPGDW LHAPLEPDVR AVVVGFDPHF SYMKLTKALR YLQQPGCLLV GTNMDNRLPL ENGRFIAGTG CLVRAVEMAA QRQADIIGKP SRFIFDCVSQ EYGINPERTV MVGDRLDTDI LLGATCGLKT ILTLTGVSTL GDVKNNQESD CVSKKKMVPD FYVDSIADLL PALQG
Tampone PGP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. .
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.