missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTMA Protein

Codice prodotto. 15992829
Click to view available options
Quantità:
1 mg
10 μg
2 μg
Dimensione della confezione:
10µg
1mg
2µg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 15992829

Marca: enQuireBio™ QP132061mg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

A cDNA sequence encoding the PTMA was constructed and used to recombinantly synthesize the protein.

Specifications

Nome PTMA Protein
Quantità 1 mg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His
Sequenza MGSSHHHHHH SSGLVPRGSH MGSMSDAAVD TSSEITTKDL KEKKEVVEEA ENGRDAPANG NANEENGEQE ADNEVDEEEE EGGEEEEEEE EGDGEEEDGD EDEEAESATG KRAAEDDEDD DVDTKKQKTD EDD
Tampone The PTMA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.