Learn More
enQuireBio™ Recombinant Human Ras association domain-containing protein 1
Codice prodotto. 16091071
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Codice prodotto. 16091071
Marca: enQuireBio™ QP7834ec100ug
A DNA sequence encoding the Homo sapiens (Human) Ras association domain-containing protein 5, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso NP_872604.1
ID gene (immissione) 83593
Nome Ras association domain-containing protein 1
Quantità 100 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene RASSF5
Lunghezza della proteina Full Length of Isoform 2
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine GST
Sequenza MTVDSSMSSG YCSLDEELED CFFTAKTTFF RNAQSKHLSK NVCKPVEETQ RPPTLQEIKQ KIDSYNTREK NCLGMKLSED GTYTGFIKVH LKLRRPVTVP AGIRPQSIYD AIKEVNLAAT TDKRTSFYLP LDAIKQLHIS STTTVSEVIQ GLLKKFMVVD NPQKFALFKR IHKDGQVLFQ KLSIADRPLY LRLLAGPDTE VLSFVLKENE TGEVEWDAFS IPELQNFLTI LEKEEQDKIQ QVQKKYDKFR QKLEEALRES QGKPG
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.