Learn More
enQuireBio™ Recombinant Human RLN2 Protein
Codice prodotto. p-7151208
Click to view available options
Quantità:
1 μg
1 mg
25 μg
5 μg
50 μg
Dimensione della confezione:
1mg
1µg
25µg
50µg
5µg
Codice prodotto. 15973199
Marca: enQuireBio™ QP13322HIS1ug
A cDNA sequence encoding the RLN2 was constructed and used to recombinantly synthesize the protein.
Specifica
Nome RLN2 Protein
Quantità 1 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His
Sequenza MGSSHHHHHH SSGLVPRGSH MGSDSWMEEV IKLCGRELVR AQIAICGMST WSKRSLSQED APQTPRPVAE IVPSFINKDT ETINMMSEFV ANLPQELKLT LSEMQPALPQ LQQHVPVLKD SSLLFEEFKK LIRNRQSEAA DSSPSELKYL GLDTHSRKKR QLYSALANKC CHVGCTKRSL ARFC
Tampone RLN2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.