missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serum amyloid P component / APCS / SAP Protein

Codice prodotto. 15926799
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 15926799

Marca: enQuireBio™ QP5659ec1mg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

A DNA sequence encoding the Homo sapiens (Human) Serum amyloid P component / APCS / SAP, was expressed in the hosts and tags indicated.

Specifications

Numero di accesso NP_001630.1
ID gene (immissione) 325
Nome Serum amyloid P component / APCS / SAP Protein
Quantità 1 mg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene APCS
Lunghezza della proteina Full Length
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine GST
Sequenza HTDLSGKVFV FPRESVTDHV NLITPLEKPL QNFTLCFRAY SDLSRAYSLF SYNTQGRDNE LLVYKERVGE YSLYIGRHKV TSKVIEKFPA PVHICVSWES SSGIAEFWIN GTPLVKKGLR QGYFVEAQPK IVLGQEQDSY GGKFDRSQSF VGEIGDLYMW DSVLPPENIL SAYQGTPLPA NILDWQALNY EIRGYVIIKP LVWV
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.