missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus
Click to view available options
Quantità:
100 μg
25 μg
500 μg
Formulazione:
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Dimensione della confezione:
100µg
25µg
500µg
Descrizione
The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus is derived from E. coli. The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus has been validated for the following applications: Bioactivity.
Specifica
Specifica
| Numero di accesso | NP_000184.1 |
| Formulazione | Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. |
| ID gene (immissione) | 6469 |
| Peso molecolare | 20 kDa |
| Nome | Sonic Hedgehog/Shh (C24II), N-Terminus |
| Quantità | 25 μg |
| Sorgente | E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 (Cys24Ile-Ile),with an N-terminal Met |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Alias gene | HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS |
| Coniugato | Unconjugated |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto