Learn More
R&D Systems™ Recombinant Human Survivin Western Blot Standard Protein
Codice prodotto. 18196580
Click to view available options
Quantità:
1 Vial
Dimensione della confezione:
100µL
Codice prodotto. 18196580
Marca: R&D Systems™ WBC010B
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
The Recombinant Human Survivin Western Blot Standard Protein has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
ID gene (immissione) 332
Nome Survivin Western Blot Standard
Quantità 1 Vial
Requisiti di stoccaggio Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Alias gene API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Coniugato Unconjugated
Recombinant Recombinant