missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX18 Protein
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Numero di accesso | NP_001073977.1 |
| ID gene (immissione) | 9096 |
| Nome | T-box transcription factor TBX18 Protein |
| Quantità | 200 μg |
| Status giuridico | Research Use Only |
| Concentrazione di endotossine | Not determined. |
| Simbolo del gene | TBX18 |
| Lunghezza della proteina | Full Length |
| Product Type | Recombinant Protein |
| Cross-reattività | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction