missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein
Click to view available options
Quantità:
25 μg
5 μg
Formulazione:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Dimensione della confezione:
25µg
5µg
Descrizione
The Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.
Specifica
Specifica
| Formulazione | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| ID gene (immissione) | 7040 |
| Peso molecolare | 12.8 kDa (monomer) |
| Nome | TGF-beta 1 |
| Quantità | 5 μg |
| Sorgente | Human embryonic kidney cell,HEK293-derived human TGF-beta 1 protein Ala279-Ser390 |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Forma strutturale | Disulfide-linked homodimer |
| Alias gene | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Coniugato | Unconjugated |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto