Learn More
Novus Biologicals™ Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein
Codice prodotto. 18269443 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
250 μg
50 μg
Dimensione della confezione:
250µg
50µg
Codice prodotto. 18269443
Marca: Novus Biologicals™ NBP261985250UG
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal hIgG-His-tag and corresponding to the amino acids 27-126 of Human TGF-beta RI/ALK-5 The Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein is derived from Baculovirus. The Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione 0.5 mg/ml
Da utilizzare con (applicazione) SDS-PAGE
Formulazione Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
ID gene (immissione) 7046
Peso molecolare 38 kDa
Quantità 250 μg
Sorgente Baculovirus
Requisiti di stoccaggio Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Status giuridico RUO
Concentrazione di endotossine < 1.0 EU per 1μg of protein (determined by LAL method)
Simbolo del gene TGFBR1
Cross-reattività TGF-beta RI/ALK-5
Specie Human
Purity or Quality Grade >90%, by SDS-PAGE