missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNF-alpha Protein
A DNA sequence encoding the Human TNF-alpha, was expressed in the hosts and tags indicated.
105.00€ - 770.00€
Specifica
Numero di accesso | NP_000585.2 |
---|---|
ID gene (immissione) | 7124 |
Nome | TNF-alpha Protein |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Less than 1.0 EU/ug as determined by LAL method. |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15923598
|
enQuireBio™
QP10352-ec-10ug |
10 μg | |||||||
15943598
|
enQuireBio™
QP10352-ec-50ug |
50 μg | |||||||
15913598
|
enQuireBio™
QP10352-ec-100ug |
100 μg | |||||||
15933598
|
enQuireBio™
QP10352-ec-250ug |
250 μg | |||||||
Specifica
NP_000585.2 | |
TNF-alpha Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MHHHHHHVRS SSRTPSDKPV AHVVANPQAE GQLQWLNRRA NALLANGVEL RDNQLVVPSE GLYLIYSQVL FKGQGCPSTH VLLTHTISRI AVSYQTKVNL LSAIKSPCQR ETPEGAEAKP WYEPIYLGGV FQLEKGDRLS AEINRPDYLD FAESGQVYFG IIAL | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
7124 | |
Research Use Only | |
TNF | |
The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg in the presence of actinomycin D. | |
Human | |
His | |
0.2 m filtered PBS, pH 7.0, lyophilized |