missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TNF RII/TNFRSF1B Protein
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
1 mg
100 μg
5 μg
500 μg
Dimensione della confezione:
100µg
1mg
500µg
5µg
Descrizione
TNF Receptor II Protein is fully biologically active when compared to standard. The ED50 as determined by its ability to inhibit the TNF-alpha mediated cytotoxicity in the L-929 cells is less than 0.2 μg/mL, corresponding to a specific activity of > 5000 IU/mg in the presence of 0.25 ng/mL of rHuTNF-alpha.
Specifica
Specifica
| Numero di accesso | P20333, P20333, P20333 |
| Da utilizzare con (applicazione) | SDS-PAGE, Bioactivity |
| Formulazione | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4. |
| ID gene (immissione) | 7133 |
| Peso molecolare | MolecularWeight-theroretical: 20.0 kDa |
| Quantità | 500 μg |
| Requisiti di stoccaggio | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Concentrazione di endotossine | Less than 0.1 EU/μg of TNF RII/TNFRSF1B as determined by LAL method. |
| Alias gene | CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, sTNF RI, sTNF RI/TNFRSF1A, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1 B, tumor necrosis factor receptor superfamily, member 1 B, Tumor necrosis factor receptor type II |
| Simbolo del gene | TNFRSF1B |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto