missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Wnt-16b Protein
Klicka för att se tillgängliga alternativ
Formulazione:
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Förpackningsstorlek:
25µg
Beskrivning
The Recombinant Human Wnt-16b Protein is derived from CHO. The Recombinant Human Wnt-16b Protein has been validated for the following applications: Bioactivity.
Specifikationer
Specifikationer
| Formulazione | Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
| ID gene (immissione) | 51384 |
| Peso molecolare | 37.6 kDa |
| Nome | Wnt-16b |
| Quantità | 25 μg |
| Sorgente | Chinese Hamster Ovary cell line,CHO-derived human Wnt-16b protein Asn30-Lys365 |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Coniugato | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 80%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering