Learn More
enQuireBio™ Recombinant Mouse EGF / Epidermal Growth Factor Protein
Codice prodotto. p-7152141
Click to view available options
Quantità:
20 μg
5 μg
Dimensione della confezione:
20µg
5µg
Codice prodotto. 15916229
Marca: enQuireBio™ QP54825ug
Recombinant Protein
Specifica
Numero di accesso NP_034243.2
ID gene (immissione) 13645
Nome EGF / Epidermal Growth Factor Protein
Quantità 5 μg
Status giuridico Research Use Only
Concentrazione di endotossine Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Simbolo del gene EGF
Attività biologica 3T3 cell proliferation significantly induced by less than 250 pg/ml of Mouse EGF / Epidermal Growth Factor (which is equivalent to activity of at least 4.0 x 10^6 units/mg or an ED50 typically around 10-100 pg/ml).
Product Type Recombinant Protein
Cross-reattività Mouse
Specie E. coli
Tag proteine Biotin
Sequenza MKKIDDDKNS . YPGCPSSYD G. YCLNGGVC MH. IESLDSY TCN. CVIGYS GDRC. QTRDL EWWEL. R
Tampone 0.5 mg/mL in sterile PBS
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.