missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CST3 k9 Protein
A cDNA sequence encoding the CST3 k9 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11543-2ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
CST3 k9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS PGAGRRGSRP GAVGGAVDAD VGEEGVQQAL DFAVREYNRA SNDAYHSRAL RVLRARKQVV SGMNYFLEVE IGRTRCTKSQ PNLDNCPFHD QPHLMRKTLC SFQIYTVPWL GKTSLVKSSC QDV | |
Purity is greater than 95% as visualized by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
K9 Cystatin-Cwas filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS, 50mM NaCl, pH 7.5. |