missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DnaK SBD Protein
A cDNA sequence encoding the DnaK SBD was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11687-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DnaK SBD Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MNEDEIQKMV RDAEANAEAD RKFEELVQTR NQGDHLLHST RKQVEEAGDK LPADDKTAIESALTALETAL KGEDKAAIEA KMQELAQVSQ KLMEIAQQQH AQQQTAGADASANNAKDDDVVDAEFEEVKDKK | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The protein contains 25mM Tris-HCl, pH 7.5, 100mM NaCl, 5mM DTT and 10%Glycerol. |