Learn More
enQuireBio™ Recombinant other DnaK SBD Protein
Codice prodotto. p-7149526
Click to view available options
Quantità:
1 mg
100 μg
20 μg
Dimensione della confezione:
100µg
1mg
20µg
Codice prodotto. 15918048
Marca: enQuireBio™ QP1168720ug
A cDNA sequence encoding the DnaK SBD was constructed and used to recombinantly synthesize the protein.
Specifica
Nome DnaK SBD Protein
Quantità 20 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Other
Specie E. coli
Tag proteine Untagged
Sequenza MNEDEIQKMV RDAEANAEAD RKFEELVQTR NQGDHLLHST RKQVEEAGDK LPADDKTAIESALTALETAL KGEDKAAIEA KMQELAQVSQ KLMEIAQQQH AQQQTAGADASANNAKDDDVVDAEFEEVKDKK
Tampone The protein contains 25mM Tris-HCl, pH 7.5, 100mM NaCl, 5mM DTT and 10%Glycerol.
Purity or Quality Grade Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.