Learn More
enQuireBio™ Recombinant other NAF Protein
Codice prodotto. 15961529
Click to view available options
Quantità:
8 mg
16 mg
32 mg
Dimensione della confezione:
16mg
32mg
8mg
Codice prodotto. 15961529
Marca: enQuireBio™ QP1278432mg
Recombinant Protein
Specifica
Nome NAF Protein
Quantità 32 mg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Other
Specie E. coli
Tag proteine Untagged
Tampone The protein (1 mg/ml) was lyophilized with no additives.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.