missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-13 Protein
Click to view available options
Formulazione:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Dimensione della confezione:
25µg
Descrizione
The Recombinant Rat IL-13 Protein is derived from E. coli. The Recombinant Rat IL-13 Protein has been validated for the following applications: Bioactivity.
Specifica
Specifica
| Formulazione | Lyophilized from a 0.2μm filtered solution in PBS. |
| ID gene (immissione) | 116553 |
| Peso molecolare | 12.3 kDa |
| Nome | IL-13 |
| Quantità | 25 μg |
| Sorgente | E. coli-derived rat IL-13 protein Thr19-His131 |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Coniugato | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto