Learn More
enQuireBio™ Recombinant S. aureus LDH-A / LDHA Protein
Codice prodotto. 16077910
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Codice prodotto. 16077910
Marca: enQuireBio™ QP7260ec1mg
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) LDH-A / LDHA, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso WP_001031880.1
ID gene (immissione) 31213039
Nome S. aureus LDH-A / LDHA Protein
Quantità 1 mg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene LDHA
Lunghezza della proteina Full Length
Product Type Recombinant Protein
Cross-reattività S. aureus
Specie E. coli
Tag proteine His-SUMO
Sequenza MNKFKGNKVV LIGNGAVGSS YAFSLVNQSI VDELVIIDLD TEKVRGDVMD LKHATPYSPT TVRVKAGEYS DCHDADLVVI CAGAAQKPGE TRLDLVSKNL KIFKSIVGEV MASKFDGIFL VATNPVDILA YATWKFSGLP KERVIGSGTI LDSARFRLLL SEAFDVAPRS VDAQIIGEHG DTELPVWSHA NIAGQPLKTL LEQRPEGKAQ IEQIFVQTRD AAYDIIQAKG ATYYGVAMGL ARITEAIFRN EDAVLTVSAL LEGEYDEEDV YIGVPAVINR NGIRNVVEIP LNDEEQSKFA HSAKTLKDIM AEAEELK
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.