Learn More
enQuireBio™ Recombinant SARS SARS Mosaic S(N) Protein
Codice prodotto. p-7151312
Click to view available options
Quantità:
1 mg
100 μg
500 μg
Dimensione della confezione:
100µg
1mg
500µg
Codice prodotto. 15903519
Marca: enQuireBio™ QP13422100ug
Recombinant Protein
Specifica
Nome SARS SARS Mosaic S(N) Protein
Quantità 100 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività SARS
Specie E. coli
Tag proteine Untagged
Tampone 25mM Tris-HCl, 0.4% sarcosyl, 0.25% Triton .–100 and 50% glycerol.
Purity or Quality Grade SARS Mosaic Protein is >95% pure as determined by 10% PAGE (coomassie staining).